Plastic Face Shield, Bal Supply
# de catalogue 76443-194
Fournisseur: Tip Industries
Spécifications
- Width:13"
- Description:Plastic Face Shield
- Height:9"
- Thickness:14 mil
- Cat. no.:76443-194
Spécifications
A propos de cet article
Premium quality face shield features a clear PET lens designed to protect the face from accidental splashes.
- PET lens, 14 mil thickness
- Anti-fog and anti-static coating
- Comfortable foam head piece
- Single-use design
- Latex-free
- Nonsterile
Premium quality, optically clear, distortion-free lens. Face shields feature a foam head piece and soft, stretchable headband for comfortable use over extended periods.
Packaging: Face shields are individually bagged, and supplied 200 per case.
Customers Who Bought This Also Bought