ContactUsLinkComponent
TDS Standard, Environmental Express
TDS Standard, Environmental Express
# de catalogue 76436-958
Fournisseur:  Environmental Express
Numéro CAS:  
Total Dissolved Solids standard, provides ongoing batch level QA/QC samples for TDS analysis at an affordable cost, Create a Lab Control Sample by transferring bottle contents into a graduated cylinder or volumetric flask, pour to volume and analyze, acceptability range of +/-3 standard deviations
TDS Standard, Environmental Express
# de catalogue 76436-958
Fournisseur:  Environmental Express
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    TDS Standard
  • Size:
    10 pk
  • Cat. no.:
    76436-958
Spécifications
A propos de cet article
Save time and money with a TDS standard compliant to SM 2540 and ASTM D5907 methods.
The Total Dissolved Solids (TDS) standard provides ongoing batch level QA/QC samples for the TDS analysis at an affordable cost. Create a Lab Control Sample (LCS) by transferring bottle contents into a graduated cylinder or volumetric flask, pour to volume and analyze. An acceptability range of ±3 standard deviations is provided for each parameter.
Each bottle of standard has a unique ID# and corresponding unique solids values.
Delivery: MSDS and Certificate of Analysis with NIST traceability and instructions for use are included with each set.
Customers Who Bought This Also Bought