SafePlus™ Maxi dispenser, Vileda Professional
# de catalogue 76437-396
Fournisseur: Vileda Professional - FHP
Spécifications
- Description:SafePlus™ Maxi dispenser for SafePlus™ maxi disposable wipes
- Cat. no.:76437-396
Spécifications
A propos de cet article
The SafePlus™ Maxi System is the safe way to dispense single-use microfiber wipes.
- Antibacterial self-closing flexible lid, never leaving the dispenser open
- Cetified by 3rd party, Hohenstein Labs, antibacterial properties on flexible lid
- Flexible lid remains antibacterial even after 80x autoclave cycles
- Sealed polybags wipes are packaged to fit perfectly in the dispensers providing extra safety in preparation, use, and storage
To achieve a high level of hygiene, the cleaning and disinfection of surfaces is becoming increasingly important. That's why Vileda Professional® offers the most hygienic and safe cleaning solutions for surface cleaning and disinfection. SafePlus™ wipes achieve the highest level of cleanliness when using a single-use microfiber wipe while cleaning and disinfecting at the same time. The SafePlus™ Maxi dispenser is one of the safest and most innovative wipe dispenser systems available in the market today.
Customers Who Bought This Also Bought