ContactUsLinkComponent
Matrix Orbital Delta Thermoshakers
Matrix Orbital Delta Thermoshakers
Fournisseur:  IKA Works
Certificats
Matrix Orbital Delta Thermoshakers
Matrix Orbital Delta Thermoshakers
Fournisseur:  IKA Works
Certificats
Spécifications
  • Catalog No:
  • 76417-516
  • 76417-514
  • 76417-520
  • 76417-518
  • Model:
  • IKA Matrix Orbital Delta F1.5
  • IKA Matrix Orbital Delta F0.5
  • IKA Matrix Orbital Delta F2.0
  • IKA Matrix Orbital Delta FP
  • Incubating accuracy:
  • ±0.5 at 20...45 °C
  • ±0.5 at 20...45 °C
  • ±0.5 at 20...45 °C
  • ±1.0 at 20...45 °C
  • Temperature control range:
  • Ambient 4…100 °C
  • Ambient 4…100 °C
  • Ambient 4…100 °C
  • Ambient 4…100 °C
  • Heating speed:
  • 11 °C/min
  • 15 °C/min
  • 13 °C/min
  • 18 °C/min
  • W×D×H:
  • 217×158×351 mm
  • 216×149×351 mm
  • 216×158×351 mm
  • 216×147×351 mm
  • Orbit:
  • 3 mm
  • 3 mm
  • 3 mm
  • 3 mm
  • Mixing speed range:
  • 0 - 1500 rpm
  • 0 - 2000 rpm
  • 0 - 1500 rpm
  • 0 - 2000 rpm