Avantor® Hichrom, Retention Gap/Precolumn, HMDS Deactivated Fused Silica
Fournisseur: VWR International
Certificats
A propos de cet article
HMDS deactivated fused silica retention gap / precolumn. Deactivation mainly for apolar solvents.
Customers Who Bought This Also Bought