Avantor® Hichrom HI-POF 2, GC Columns
Fournisseur: VWR International
Certificats
A propos de cet article
Application specifc columns developed for pesticides, herbicides, insecticides analysis and so on.
Maximum temperature limits may change depending on the film thickness.
Spécifications
- Catalog No:
- 76428-198
- 76428-200
- 76428-194
- 76428-196
- 76428-212
- 76428-214
- 76428-208
- 76428-210
- 76428-206
- 76428-202
- 76428-204
- Phase name:
- HI-POF 2
- HI-POF 2
- HI-POF 2
- HI-POF 2
- HI-POF 2
- HI-POF 2
- HI-POF 2
- HI-POF 2
- HI-POF 2
- HI-POF 2
- HI-POF 2
- Phase composition:
- Proprietary Phase developed for pesticides
- Proprietary Phase developed for pesticides
- Proprietary Phase developed for pesticides
- Proprietary Phase developed for pesticides
- Proprietary Phase developed for pesticides
- Proprietary Phase developed for pesticides
- Proprietary Phase developed for pesticides
- Proprietary Phase developed for pesticides
- Proprietary Phase developed for pesticides
- Proprietary Phase developed for pesticides
- Proprietary Phase developed for pesticides
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes