Avantor® Hichrom HI-35 HT, GC Columns
Fournisseur: VWR International
Certificats
A propos de cet article
General purpose column with added high temperature stability for the analysis of Pesticides, PCBs, substituted polar compounds, phenols, etc.
- Ideal as confrmation column
Max temperature limits may change depending on the film thickness.
Spécifications
- Catalog No:
- 76431-598
- 76431-618
- 76431-816
- 76431-596
- 76431-594
- 76431-814
- 76431-616
- 76431-614
- 76431-592
- 76431-612
- 76431-590
- 76431-588
- 76431-610
- 76431-586
- 76431-608
- 76431-606
- 76431-584
- 76431-604
- 76431-582
- 76431-602
- 76431-600
- 76431-622
- 76431-620
- Phase name:
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- HI-35 HT
- Phase composition:
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- 35% Phenyl, 65% Methyl Polysiloxane - high temperature
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Crossbond:
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Temperature limit:
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- USP code:
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42