Avantor® Hichrom HI-5 HT, GC Columns
Fournisseur: VWR International
Certificats
A propos de cet article
General purpose column with added temperature stability. For the analysis of solvent impurities, PCBs, hydrocarbons, essential oils, semivolatiles, pesticides, phenols and more.
Max temperature limits may change depending on the film thickness.
Spécifications
- Catalog No:
- 76431-658
- 76431-656
- 76431-654
- 76431-652
- 76431-650
- 76431-648
- 76431-646
- 76431-644
- 76431-642
- 76431-640
- 76431-186
- 76431-818
- 76431-638
- 76431-636
- 76431-634
- 76431-632
- 76431-630
- 76431-628
- 76431-626
- 76431-624
- 76431-820
- 76431-664
- 76431-662
- 76431-660
- Phase name:
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- Phase composition:
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature limit:
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- USP code:
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41