Avantor® Hichrom HI-SOLVE 1, GC Columns
Fournisseur: VWR International
Certificats
A propos de cet article
Application specifc columns developed for complex solvents mixtures analysis. HI-SOLVE 1 is ideal for aromatics and oxygenates in gasoline.
Max temperature limits may change depending on the film thickness.
Spécifications
- Catalog No:
- 76429-096
- Phase name:
- HI-SOLVE 1
- Phase composition:
- Proprietary Phase developed for complex solvent mix
- Crossbond:
- Stabilized