Avantor® Hichrom HI-50, GC Columns
Fournisseur: VWR International
Certificats
A propos de cet article
High polarity phase, ideal for detailed FAMEs and Dioxins isomers separation.
- Phase composition: 50% Cyanopropyl, 50% Methyl Polysiloxane
Max temperature limits may change depending on the film thickness.
Spécifications
- Catalog No:
- 76430-452
- 76425-864
- 76425-862
- 76425-906
- 76425-866
- 76425-900
- 76425-898
- 76425-860
- 76425-904
- 76425-902
- 76425-892
- 76425-890
- 76425-896
- 76425-894
- 76425-888
- 76425-886
- 76430-448
- 76425-884
- 76430-450
- 76430-444
- 76430-446
- 76425-878
- 76425-876
- 76425-920
- 76425-882
- 76425-880
- 76425-870
- 76425-914
- 76425-868
- 76425-912
- 76425-874
- 76425-918
- 76425-872
- 76425-916
- 76425-910
- 76425-908
- Phase name:
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- Phase composition:
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- Polarity:
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- Crossbond:
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Stabilized
- Temperature limit:
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- USP code:
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8