Avantor® Hichrom HI-OVW, GC Columns
Fournisseur: VWR International
Certificats
A propos de cet article
Max temperature limits may change depending on the film thickness.
Spécifications
- Catalog No:
- 76427-126
- Phase name:
- HI-OVW
- Phase composition:
- Proprietary Phase developed for Waxes Analysis in Edible Oils
- Crossbond:
- Yes