Avantor® Hichrom HI-VOC 1, GC Columns
Supplier: VWR International
Certificates
About this item
Application specifc columns for volatile organic compounds (VOCs), solvents and purgeable compounds. Due to the high max. temperature, they are ideal for two-parallel columns confg. in the same oven. A good alternative to the HI-624 giving different selectivity. It is a more robust than the HI-624 and can operate at higher temperatures.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76429-192
- 76429-194
- 76429-188
- 76429-190
- 76429-118
- 76429-184
- 76429-186
- 76429-114
- 76429-158
- 76429-180
- 76429-116
- 76429-182
- 76429-154
- 76429-112
- 76429-156
- 76429-128
- 76429-172
- 76429-130
- 76429-174
- 76429-124
- 76429-168
- 76429-126
- 76429-170
- 76429-120
- 76429-164
- 76429-122
- 76429-166
- 76429-160
- 76429-162
- 76429-136
- 76429-138
- 76429-132
- 76429-176
- 76429-134
- 76429-178
- 76429-150
- 76429-152
- 76429-146
- 76429-148
- 76429-142
- 76429-144
- 76429-140
- Phase name:
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- Phase composition:
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes