Avantor® Hichrom HI-1 HT, GC Columns
Fournisseur: VWR International
Certificats
A propos de cet article
General purpose GC-MS column Solvent impurities, PCBs, SimDist, drugs, natural gases, essential oils, semivolatiles, pesticides, phenols, etc.
Max temperature limits may change depending on the film thickness.
Spécifications
- Catalog No:
- 76431-458
- 76431-456
- 76431-494
- 76431-496
- 76431-490
- 76431-492
- 76431-486
- 76431-488
- 76431-460
- 76431-462
- 76431-804
- 76431-802
- 76431-476
- 76431-478
- 76431-472
- 76431-474
- 76431-468
- 76431-470
- 76431-464
- 76431-466
- 76431-482
- 76431-484
- 76431-480
- 76431-122
- Phase name:
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- Phase composition:
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- 100% methyl polysiloxane (100% dimethylpolysiloxane) - high temperature
- Polarity:
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature limit:
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- USP code:
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38