Avantor® Hichrom HI-BIODIESEL 110, GC Columns
Fournisseur: VWR International
Certificats
A propos de cet article
Application specifc column for free and total glycerine (phase stable up to 380 °C) and for FAMEs in biodiesel analysis.
Max temperature limits may change depending on the film thickness.
Spécifications
- Catalog No:
- 76426-560
- 76426-558
- Phase name:
- HI-BIODIESEL 110
- HI-BIODIESEL 110
- Phase composition:
- Proprietary Phase developed for Biodiesel EN14110 analysis
- Proprietary Phase developed for Biodiesel EN14110 analysis
- Crossbond:
- Yes
- Yes
- Temperature limit:
- 380 °C
- 380 °C