Assurance® Grade ICP Standards Kit, 38 Single-Element Standards, SPEX CertiPrep
# de catalogue SXICP-KIT-1
Fournisseur: SPEX CERTIPREP LLC
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:LDPE bottle
- Description:ICP Standards Kit, 38 Single-Element Standards
- Size:38x 125 ml
- Cat. no.:SXICP-KIT-1
Spécifications
A propos de cet article
Assurance® Grade, set of 38 single-element standards
Customers Who Bought This Also Bought