ContactUsLinkComponent
Assurance® Grade Mixed Calibration Standards, Set of 5, for AA and ICP, SPEX CertiPrep
Assurance® Grade Mixed Calibration Standards, Set of 5, for AA and ICP, SPEX CertiPrep
# de catalogue SXMIXSTD-SET
Fournisseur:  SPEX CERTIPREP LLC
Numéro CAS:  
Assurance® Grade Mixed Calibration Standards, Set of 5, for AA and ICP, SPEX CertiPrep
# de catalogue SXMIXSTD-SET
Fournisseur:  SPEX CERTIPREP LLC
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Packaging:
    LDPE bottle
  • Description:
    Mixed Calibration Standards, Set of 5
  • Size:
    5x 125 ml
  • Cat. no.:
    SXMIXSTD-SET
Spécifications
A propos de cet article
Assurance® mixed multi-element standards for AA & ICP.
Standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only SPEX CertiPrep Matrix Blanks when diluting your standards.
AA & ICP Calibration Standards are provided for routine instrument calibration. The concentrations and matrices have been selected for convenience of use and stability.
For use in US EPA Method 200.7 (Revision 4.4) and SW-846, Method 6010 (Third Edition).
Customers Who Bought This Also Bought