ContactUsLinkComponent
Assurance® Grade Interference Check Multi-Element Standards with Mercury for AA & ICP, SPEX CertiPrep
Assurance® Grade Interference Check Multi-Element Standards with Mercury for AA & ICP, SPEX CertiPrep
# de catalogue SXINTER-SET
Fournisseur:  SPEX CERTIPREP LLC
Numéro CAS:  
Assurance® Grade Interference Check Multi-Element Standards with Mercury for AA & ICP, SPEX CertiPrep
# de catalogue SXINTER-SET
Fournisseur:  SPEX CERTIPREP LLC
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Packaging:
    LDPE bottle
  • Description:
    Interference Check Multi-Element Standards with Mercury
  • Size:
    3x 125 ml
  • Cat. no.:
    SXINTER-SET
Spécifications
A propos de cet article
Assurance® mixed multi-element interference check standards for AA & ICP.
Standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only SPEX CertiPrep Matrix Blanks when diluting your standards.
The Interference Check Standards are used to set or confirm that the correct background correction intervals have been set for sequential ICP spectrometers and that the proper inter-element correction factors are set for simultaneous ICP spectrometers.
For use in US EPA Method 200.7 (Revision 4.4) and SW-846, Method 6010 (Third Edition).
Customers Who Bought This Also Bought