Edge® 48-919 Light-Duty Multi-Purpose Industrial Gloves, Ansell
Fournisseur: Ansell Healthcare
Certificats
A propos de cet article
Cost effective solution for lightweight applications requiring a durable coating and secure grip in oily environments.
- Excellent abrasion resistance
- Double layer nitrile coating keeps hands dry and comfortable when handling oily parts and tools
- Elastic knit-wrist ensures snug fit
- Applicable in oily environments
Made of a seamless liner, fully dipped into NBR coating. The gloves are fully NBR dipped to provide protection against industrial fluid exposure. The rough palm coating ensures high secure grip in oily environment. Designed for workers who handle small and medium-sized components coated with oil or lubricant. Use primarily for automotive, construction, machinery/equipment, and metal fabrication applications.
Certifications: EN 388:2016. Category II.
Packaging: Gloves are packaged 12 pairs per bag, 12 bags per carton.
Spécifications
- Catalog No:
- 76424-294
- 76424-292
- 76424-290
- 76424-288
- 76424-286
- 76424-284
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Gauge:
- 13
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist