VWR® 96-Well EVA and Silicone Microplate Covers
Fournisseur: VWR International
Certificats
A propos de cet article
VWR® 96-well covers are designed for chromatography and high throughput screening (HTS) applications.
- Covers for chromatography and storage
- Ultra-pure silicone
- SBS and ANSI standard footprint
- Teflon barrier eliminates cross contamination of samples
- 80 °C dry heat autoclavable
- Resists coring and tearing
VWR® 96-well covers are manufactured specifically for chromatography. The multiple options ensure sample security, no matter the application. Non-cut and pre-cut versions cover the majority of all instrument requirements.
Customers Who Bought This Also Bought