SP Bel-Art® Sterileware® Powder Probe Samplers, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Ideal for taking a cross sectional sample through a bulk powder, this probe is available in two convenient lengths.
- End tip contains free-flowing powders within the probe
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) probe is heat sealed in individual polyethylene bag
- Gamma irradiated sterile (SAL 10⁻⁶)
- Ready for one time use and disposal
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Used by pharmaceutical, food, cosmetic and chemical companies.
Ordering information: Sterilized products are not returnable.
Customers Who Bought This Also Bought