SP Bel-Art® Sterileware® Powder Lance Samplers, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
A simple insert, rotate, and withdraw motion allows cross sectional sampling of non-flowing, bulk powders.
- Open blade works well with sticky, cohesive powders
- Rounded edges help hold sample in place
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) lance is heat sealed in individual polyethylene bag
- Gamma irradiated sterile (SAL 10⁻⁶)
- Ready for one time use and disposal
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Available in three lengths.
Ordering information: Sterilized products are not returnable.
Customers Who Bought This Also Bought