High Volume MSL 8 Digital Magnetic Stirrers, Velp Scientific
Fournisseur: VELP SCIENTIFIC INC.
Certificats
A propos de cet article
Digital magnetic stirrer provides continuous operations both at low and high speed, with timer, auto-reverse and SpeedServo.
- SpeedServo™ technology maintains constant speed
- Autoreverse of the stirring direction (5 s to 99 min 55 s)
- Intermittent mode programs to meet workflows
- Resistant stainless steel cover and epoxy painted metal structure
- Safety Lock feature to avoid possible alteration of the working conditions
Gentle start to protect your sample. The control panel is protected from liquid spills. The brushless motor provides continuous operation for high volumes at any speed. The clear LCD display ensure accurate speed setting of 10 rpm.
Automatic shut dowm display after 10 s.
Delivery: Three year warranty.
Spécifications
- Catalog No:
- 76416-226
- 76416-224
- 76416-222
- Model:
- MSL 50 digital
- MSL 25 digital
- MSL 8 digital
- Plate material:
- Stainless steel
- Stainless steel
- Stainless steel
- Max. stirring capacity H₂O:
- 50 L
- 25 L
- 8 L
- Timer:
- 1 min - 99 h 59 min
- 1 min - 99 h 59 min
- 1 min - 99 h 59 min