ARCH Probes for the Opt-Diss, Distek
Fournisseur: DISTEK, INC.
Certificats
A propos de cet article
Patented Fiber optic ARCH Probes for use with Opt-Diss 405 and 410 UV Fiber optic system.
- Variety of pathlengths
- Negligible hydrodynamic effect
- Patented design
Designed for dissolution testing.
Customers Who Bought This Also Bought