Bel-Art Sterileware® Double Bagged Long Handle Sampling Spoons, SP Scienceware
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Minimizing contamination of small sample collection. Ten individually packaged scoops are sealed in an outer bag to create multiples layers of protection against contaminants and debris.
- Ideal for use in multi-zone clean rooms
- FDA grade polystyrene safe for food, drug and cosmetic use
- Gamma irradiated sterile to SAL 10⁻⁶
- Lot stamped for accurate lot tracing, and shipped with a certificate of processing for sterilization
The long handle on these spoons enables users to reach into jars, bottles and other confined areas to minimize the potential of personal contact with sampled material.
Customers Who Bought This Also Bought