ContactUsLinkComponent
QuantiQuik™ Oxalate Quick Test Strips, BioAssay Systems
QuantiQuik™ Oxalate Quick Test Strips, BioAssay Systems
# de catalogue 76423-322
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Oxalate Quick Test Strip
QuantiQuik™ Oxalate Quick Test Strips, BioAssay Systems
# de catalogue 76423-322
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Oxalate quick test strips
  • Cat. no.:
    76423-322
  • No. of Tests:
    10 tests
Spécifications
A propos de cet article
Direct determination of oxalate concentrations in biological samples.
  • Fast and sensitive: use of 10 µl sample, semi-quantitative measurement between 0 to 1.5 mg/dl or 0 to 170 µm (undiluted) oxalate
  • Method: visual
  • Samples: serum, plasma, urine, and more
Oxalate, or oxalic acid, is a metabolic breakdown product of the Kreb's cycle in eukaryotes and the glyoxylate cycle in other microorganisms. Typically present in the urine of humans and other mammals, oxalate concentration can be used as a measure of kidney function. A high level of oxalate is an indicator for kidney stones, which are primarily made of the insoluble salt calcium oxalate. Measuring oxalate is more accurate than measuring calcium as a marker for kidney stones because calcium is excreted at high concentrations even in normal urine.
BioAssay Systems QuantiQuik™ oxalate quick test strips are based on the enzymatic oxidation of oxalate coupled with a color reaction in one step. The intensity of product color is directly proportional to the oxalate concentration in the sample.
Customers Who Bought This Also Bought