Desktop Thermal Transfer Label Ribbons, UltraTape
Fournisseur: Ultratape Industries
Certificats
A propos de cet article
Printer ribbons designed for high performance printing on thermal transfer labels.
- All-purpose
- Cleanroom compatible
- High temperature
- Chemical-resistant
Thermal transfer ribbons are designed for use specifically with the UltraLabel PRO™ Cleanroom printer and UltraLabel Pro-600 cleanroom printer series.
Customers Who Bought This Also Bought