PIG® PR70 Heavy-Duty Maintenance Wipers, New Pig
Catalog # 89069-048
Supplier: New Pig
Specifications
- Color:White
- Description:PR70 heavy-duty wipers
- Cat. no.:89069-048
- Packaging:Jumbo roll
- W×L:25.4×30.5 cm (10×12")
- Supplier No.:WIP309
Specifications
About this item
This strong, absorbent, low-lint roll is ideal for daily heavy-duty jobs.
- Strong and solvent-resistant for wiping machines and rough parts, but soft enough for cleaning skin or preparing surfaces
- Exceptional abrasion resistance and wet strength that stands up to rough surfaces, jagged edges, or metal chips
- Excellent for heavy-duty maintenance jobs like machine wipedowns and degreasing parts
- Economical jumbo rolls dispense easily and offer a lower cost per wiper for high-volume use
- Low-lint construction won't shed, pill, or leave behind unwanted fibers when wiping parts
Binder-free construction withstands solvents, degreasers, and cleaning agents.
Customers Who Bought This Also Bought