Metro® Sanitizer Holder for Super Erecta® Wire Shelving and SmartWall™ Shelving, Intermetro Industries
Fournisseur: Metropolitan Wire
Certificats
A propos de cet article
Conveniently store sanitizer on Metro Super Erecta® and SmartWall™ shelving systems with the sanitizer holder.
- One gallon holder fits bottles up to 6.25" diameter
- Two liter holder fits bottles up to 4.75" diameter
All holders are supplied with an optional retaining loop. Sanitizer is not included.
Customers Who Bought This Also Bought