Table-Gard Disposable Work Mats, Blue Flowers, Accuform
Supplier: ACCUFORM MANUFACTURING, INC
Certificates
About this item
Create safe, clean workspaces by applying the adhesive mat to surfaces that are not regularly cleaned.
- Provides protection from germs, dirt, and chemical residue
- Compact and lightweight mat fits in workbags for on-the-go working needs
- Made of a smooth, thin 2-mil plastic film that is waterproof and tear-resistant
- Can be discarded in waste or recycle bin (recycle code 5)
- 12.5×19.5" size creates a workspace large enough for laptops and folios
- BPA, Latex, and PVC-free material without toxic chemicals
- The adhesive is safe for indirect food contact (complies with FDA 21 CFR 175.105)
The self-adhesive mat is repositionable and lifts easily for disposal when work is complete. The mat is safe for application to countertops, desktops, conference tables, and restaurant tables.
Customers Who Bought This Also Bought