Anti-Static Cleanroom Tapes with Conductive Adhesive, UltraTape
Fournisseur: Ultratape Industries
Certificats
A propos de cet article
Particle-free, residue-free tapes.
Specially constructed self adhesive tape that is based on a polyester film that has been coated with an anti-static primer, plus an anti-static pressure sensitive acrylic adhesive layer.
Customers Who Bought This Also Bought