Corepoint Scientific™ General Purpose Series Stainless Steel Freezer, Horizon Scientific
Catalog # 76422-118
Supplier: Horizon Scientific
Specifications
- Capacity:651 L (23 cu.ft.)
- Description:General purpose series stainless steel freezer, GPF231SSS/0A3
- Environmentally Preferable:Safer Human and Environmental Health,Low Manufacturing Impact,Energy Efficient,Waste Reducing,Water Efficient
- Model:GPF231SSS/0A3
- Environmental certification:Natural refrigerants
- Cat. No.:76422-118
- Defrost:Automatic
- Electrical:115 V, 60 Hz, 8 A, 3/4 HP
- Shelves/drawers:3 adjustable
- Door type:Solid door
- Shipping weight:148 kg (327 lb.)
- Exterior W×D×H:686×864×2115 mm (27×34×83¼")
Specifications
About this item
General purpose solid door stainless steel freezer with digital display microprocessor temperature controller. This unit features hydrocarbon natural refrigerants and sensor error alarm and remote alarm contacts. Audible and visual high/low alarms and forced draft circulation are some of the features of this well rounded unit. Operating temperature range to –30 °C.
- Natural refrigerant
- Keyed door lock
- Probe access port
- Integrated door handle
Certifications: ETL listed
Caution: Auto defrost freezers incorporate an electric heater installed on the evaporator that assists in the removal of the frost and ice. Although the heater helps speed up the frost and ice removal, it can raise the temperatures within the freezer.
Customers Who Bought This Also Bought