Vinyl Medium Adhesion Cleanroom Tapes, UltraTape
Fournisseur: Ultratape Industries
Certificats
A propos de cet article
Particle-free, residue-free tapes.
- Chemical-resistant
- Easily marked with a pen
- Wide assortment of colors and sizes
General use tape with medium adhesion provides a strong bond and easy release, making it perfect for mainteance and construction uses. The tape is chemical-resistant, available in an assortment of colors, and can be written on with pen.
Customers Who Bought This Also Bought