ContactUsLinkComponent
Ward’s Nylon Rope Trick and Synthesis Kit
Ward’s Nylon Rope Trick and Synthesis Kit
# de catalogue 470328-906
Fournisseur:  VWR International
Ward’s Nylon Rope Trick and Synthesis Kit
# de catalogue 470328-906
Fournisseur:  VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Nylon Rope Trick
  • Cat. no.:
    470328-906
Spécifications
A propos de cet article
Impress your students with this fun demonstration.
  • Show an example of polymer chemistry from two colorless solutions
  • Engage students in polymer chemistry
  • Teach real-world applications of organic chemistry
Perform the popular nylon rope trick/demonstration with this fun and easy kit. Nylon, a common man-made fiber, is actually a synthetic polymer. This kit is designed to demonstrate the principles of polymeric bonding and the formation of nylon by a condensation polymerization reaction. Instructions include steps to prepare Nylon 6-10 with the Nylon Rope Trick and Nylon 6-6 with Nylon synthesis. Engage students in discussions revolving around different types of polymers in the fields of chemistry and biology. The demonstration and synthesis requires approximately 15 minutes.
Customers Who Bought This Also Bought