Advanced Vortex Mixers, ZX4, VELP Scientifica
Supplier: VELP SCIENTIFIC INC.
Certificates
About this item
Vortex mixer with adjustable stirring speed, with continuous and IR sensor operating mode.
- Variable speed operation (electronically controlled)
- Large choice of accessories to suit tubes, plates and flasks
- Universal voltage/frequency model
- Accessories fit by simple pressure
- Four large, anti-slip rubber feet for staying in required position
Optimum resistance to chemicals from technopolymer casing, excellent stability with zinc alloy base.
The infrared sensor technology ensures a drastic reduction in repetitive strain.
Certifications: IP Protection Class according to CEI EN 60259: IP 42 protection against particles and liquids.
Delivery: Three year warranty.
Supplied with small rubber supporting plate, customizable soft foam top and stand are available as options.
Supplied with small rubber supporting plate, customizable soft foam top and stand are available as options.
Specifications
- Catalog No:
- 76416-206
- 76416-292
- Orbit:
- 4.5 mm
- 4.5 mm
- Input power:
- 15 W
- 15 W