RH Basic and Digital Magnetic Hotplate Strirrers, IKA
Fournisseur: IKA Works
Certificats
A propos de cet article
Magnetic hotplate stirrers with stainless steel heating plate, white units have an enamelled plate. All models are easy to operate and have adjustable safety temperature from 50 to 370 °C. High heat output of 600 watts and steel-copper compound hot plate ensures rapid heating.
- RH basic includes M 10 thread for a support rod
- Soft-start of the stirring motor for both models
- Strong magnetic field and wide speed range for volumes up to 15 litres
- RH digital has digital display of temperature and speed
- Error code display and touch keypad (RH digital)
- Speed display scale (basic) and LED (digital)
The RH digital model has bushing according to DIN 12878 for connecting an electronic contact thermomete. Highly accurate temperature control is possible with sensor placed directly in the medium.
Maximum stirring bar length of 80 mm, Heating rate of 6 K/min.
Certifications: IP protection class according to DIN EN 60529: IP 21
Spécifications
- Catalog No:
- 76414-086
- 76414-084
- 76414-082
- 76414-080
- Model:
- RH digital white
- RH digital
- RH basic white
- RH basic
- Plate dimensions:
- Ø 135 mm
- Ø 135 mm
- Ø 135 mm
- Ø 135 mm
- Max. stirring capacity H₂O:
- 15 L
- 15 L
- 15 L
- 15 L
- Speed range:
- 50 - 2000 rpm
- 50 - 2000 rpm
- 50 - 2000 rpm
- 50 - 2000 rpm
- W×D×H:
- 160×246×90 mm
- 160×246×90 mm
- 160×246×90 mm
- 160×246×90 mm
- Temperature control accuracy:
- ETS-D6 : ±0.2 K
- ETS-D5 : ±0.5 K
- ETS-D6 : ±0.2 K
- ETS-D5 : ±0.5 K
- Heat output:
- 600 W
- 600 W
- 600 W
- 600 W