Universal Hand Sanitizer Stands and Holders, Marlin Steel Wire Products
Fournisseur: Marlin Steel Wire Products
Certificats
A propos de cet article
A custom hand hygiene solution for ensuring sanitation in any facility. The plain steel, black-coated stand is designed to work with different dispenser models across various brands, as well as Marlin’s no-touch sanitizer dispenser.
- Varied models are designed to hold specific sanitizer dispensing products
- Universal stand works with SC Johnson, Deb, Purell, and Martin Steel brands
- Portable stands can be easily moved to convenient locations or areas of high traffic
The universal stand is designed for the demanding applications of manufacturing, food processing, medical industries, and more, all while ensuring that employees can stay safe and productive. Installing hand sanitizer stations will minimize risks of COVID-19 transmission and other diseases spreading between coworkers. Hand sanitizer stations are a convenient way to ensure sterile conditions and workplace safety since they can be set up wherever is most accessible for employees.
Ordering information: Dispenser is not included unless specified.
Customers Who Bought This Also Bought