Heratherm® Drying Ovens, General Protocol Series, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Heratherm® General Protocol ovens which provide an ideal heating and drying solution for research, clinical or industrial needs.
- Operating temperatures from 50 to 250 °C
- Automatic over-temperature alarm system
- Low overall energy consumption
- Built in timer
- Large, easy to read vacuum fluorescent display
- Doors can be opened at over 180 °C
Easy to use, microprocessor-controlled touch button operation. Corrosion-resistant stainless steel chambers with rounded corners.
Delivery: Two year warranty.
Spécifications
- Catalog No:
- 76416-326
- 76416-330
- 76416-328
- 76416-334
- 76416-332
- 76416-336
- Temperature stability:
- ±0.4 °C
- ±0.4 °C
- ±0.4 °C
- ±0.4 °C
- ±0.3 °C
- ±0.3 °C
- Control type:
- Microprocessor control with vacuum fluorescent display
- Microprocessor control with vacuum fluorescent display
- Microprocessor control with vacuum fluorescent display
- Microprocessor control with vacuum fluorescent display
- Microprocessor control with vacuum fluorescent display
- Microprocessor control with vacuum fluorescent display
- Timer:
- On/off
- On/off
- On/off
- On/off
- On/off
- On/off