ContactUsLinkComponent
NPS® Elephant Z-Stool, Black Seat and Chrome Frame, 18", National Public Seating
NPS® Elephant Z-Stool, Black Seat and Chrome Frame, 18", National Public Seating
# de catalogue 76276-936
NPS® Elephant Z-Stool, Black Seat and Chrome Frame, 18", National Public Seating
# de catalogue 76276-936
Spécifications
  • Depth:
    16.25"
  • Width:
    16.75"
  • Description:
    Stool
  • Height:
    18"
  • Model:
    76276-936
  • Weight capacity:
    300 lbs.
  • Cat. no.:
    76276-936
  • Weight:
    9.5 lbs.
Spécifications
A propos de cet article
This stool is the zenith of ergonomic design and comfort, perfect for art studios or science labs.
  • 14-gauge chrome-plated eye-shaped tubular steel frame
  • Ergonomic blow molded contoured seat providing maximum air comfort
  • Nests on tabletops for easy cleanup and storage
  • 300 lbs. weight capacity
Strong as an elephant and quiet as a mouse! The gorgeous 6600 Series Elephant Z-Stool is the zenith of ergonomic design and stylish comfort, perfect for hands-on settings like art studios or science labs. The mammoth, chrome-plated, 14-gauge, eye-shaped tubular steel frame is available in three separate height options. Each stool features a black, lightweight, blow-molded, polypropylene seat, contoured to fit the user.
Sturdy, U-shaped, sled-base legs distribute weight across a wider surface, minimizing pressure on floors. The glides are securely bonded with thermoplastic polyurethane (TPU) and move softly and smoothly over any surface. Z-Stools are conveniently stackable (six units high) or hang with ease from tabletop surfaces, allowing for simple clean-up and storage.
Certifications: MAS certified Green, ANSI-BIFMA certified.
Ordering information: 48 hours Quickship. 10 year warranty.
Customers Who Bought This Also Bought