A propos de cet article
Versatile and efficient glassware washers designed for efficient and consistent washing of narrow-neck glassware and general labware in critical washing environments.
- Touch screen display with intuitive CleanWorks OS software
- Flexible design allows for use of spindle injection and standard racks
- Maximum wash temperature 93 °C removes tough contaminants
- Conductivity sensor and HEPA-filtered forced air spindle drying with SpeedVent technology
- Durable Type 304 stainless steel chamber and racks and detergent storage cabinet
- 36-place Spindle Injection Rack included
- Liquid detergent and acid neutralizing rinse aid pumps included
These glassware washers are designed for efficient and consistent washing of narrow-neck glassware and general purpose labware in critical environments such as pharmaceutical laboratories. The CleanWorks OS software is accessible through a full-color touch screen interface and provides intuitive and clear monitoring of wash loads – ensuring repeatable, effective and traceable cleaning. Vantage models reach a maximum of 93 °C wash temperature and monitor water cleanliness with a standard conductivity sensor.
FlaskScrubber Vantage models utilize HEPA filtered forced air spindle injection and SpeedVent heat-management technology to quickly dry labware. All Vantage models include a 36-place lower spindle rack, dispensing pumps for liquid detergent and acid neutralizing rinse, and a batch viewing window. Vantage models may be outfitted with a wide variety inserts and additional racks to accommodate all types of labware.
Spécifications
- Catalog No:
- 76420-734
- 76420-732
- Environmentally Preferable:
- Waste Reducing, Water Efficient
- Waste Reducing, Water Efficient