A propos de cet article
Versatile and efficient glassware washers that can handle a wide range of wide-mouth beakers and general purpose labware.
- Touch screen display with intuitive CleanWorks OS software
- Flexible design allows for use of standard and injector spindle racks
- Maximum wash temperature of 93 °C removes tough contaminants
- Advanced wash pump maximizes water circulation up to 112 gal/min (424 liters/min)
- Standard forced air chamber drying with SpeedVent technology
- Durable Type 304 stainless steel chamber and racks
These are designed for efficient and consistent washing of general purpose labware such as wide-mouth beakers. The CleanWorks OS software is accessible through a full-color touch screen interface and provides intuitive and clear monitoring of wash loads – ensuring repeatable, effective and traceable cleaning.
SteamScrubber models reach a maximum of 93 °C wash temperature and quickly dry glassware with standard forced air chamber drying with SpeedVent heat-management technology. These models include lower and upper standard racks to accommodate a wide variety of racks and inserts making these washers extremely versatile for all washing needs.
Spécifications
- Catalog No:
- 76420-698
- 76420-702
- 76420-700
- 76420-684
- 76420-706
- 76420-704
- 76420-688
- 76420-686
- 76420-692
- 76420-690
- 76420-696
- 76420-694
- Model:
- SteamScrubber® free standing
- SteamScrubber® free standing
- SteamScrubber® free standing
- SteamScrubber® free standing
- Environmentally Preferable:
- Waste Reducing, Water Efficient
- Waste Reducing, Water Efficient
- Waste Reducing, Water Efficient
- Waste Reducing, Water Efficient
- Waste Reducing, Water Efficient
- Waste Reducing, Water Efficient
- Waste Reducing, Water Efficient
- Waste Reducing, Water Efficient
- Waste Reducing, Water Efficient
- Waste Reducing, Water Efficient
- Waste Reducing, Water Efficient
- Waste Reducing, Water Efficient