Sartolab® P20 and P20 Plus Pressure Filtration Units, Sartorius
Fournisseur: Sartorius
Certificats
A propos de cet article
Ready to use units which, attached to a membrane pump or tube pump, quickly and reliably sterilise 100 ml to 5 litres of media and aqueous solutions.
- Air vent with PTFE membrane
Cellulose acetate/PES membrane, P20 Plus with glass fibre prefilter; PC housing, sterile.
Outlet: Hose nipple with filling bell.
Non-toxic as confirmed with L929 fibrolast cells of mice and with MRC-5 lung cells of human embryonic origin.
Certifications: Pass the USP Plastics-Class VI-Test.
Packaging: Individually sterile bagged.
Spécifications
- Catalog No:
- 76420-746
- 76420-744
- 76420-748
- Water flow rate:
- 1000 ml/min (1 bar)
- 400 ml/min (1 bar)
- 400 ml/min (1 bar)
- Max. pressure:
- 3 bar
- 3 bar
- 3 bar