Data Printer for Sartorius Balances, Sartorius
# de catalogue CA97021-988
Fournisseur: Sartorius
Spécifications
- Description:Data printer
- Cat. no.:CA97021-988
- Height:21.5 cm
- Weight:0.9 kg
Spécifications
A propos de cet article
This data printer suitable for external verification with date, time, statistics, numerator.
- Values transferred from an on-line weighing instrument are stored
In addition to recording individual weighing data, the data printer can generate a statistical evaluation of weighing data.
Certifications: This data printer meets the quality standards of the chemical and pharmaceutical industries, in compliance with the requirements of GLP (Good Laboratory Practice) and GMP (Good Manufacturing Practice).
Delivery: Includes cable.
Customers Who Bought This Also Bought