ContactUsLinkComponent
Viral Transport Medium (VTM), Hardy Diagnostics
Viral Transport Medium (VTM), Hardy Diagnostics
# de catalogue 76334-640
Fournisseur:  Hardy Diagnostics
Numéro CAS:  
Viral Transport Media, 3ml, Dimension: 15x120mm Centrifuge Tube, pack of 20
Viral Transport Medium (VTM), Hardy Diagnostics
# de catalogue 76334-640
Fournisseur:  Hardy Diagnostics
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Viral transport medium
  • Size:
    3 ml
  • Cat. no.:
    76334-640
Spécifications
A propos de cet article
Hardy Diagnostics Viral Transport Medium is recommended for the collection and transport of clinical specimens for the recovery of viral agents.
  • Selective agents are added to inhibit bacterial and fungal contaminants
  • Medium acts as a cryoprotectant to ensure virus stabilization through freezing and thawing
Hardy Diagnostics Viral Transport Medium is recommended for the collection and transport of clinical specimens for the recovery of viral agents including, but not limited to, Herpes Simplex Type I, Herpes Simplex Type II, Cytomegalovirus (CMV), Influenza A, Influenza B, Respiratory Syncytial Virus (RSV), Echovirus, Adenovirus, etc.
While the optimal long term storage temperature has been determined to be 2 to 8 °C due to the COVID-19 pandemic, recent internal studies have supported that the media can be stored at room temperature (RT) without decreased performance. Data permitting, the length of RT storage shall continue to be extended as the real-time findings are collected, and applied to each lot. For the length of allowable RT storage (post retrieval from 2 to 8 °C conditions), please refer to the respective Certificate of Analysis for the lot-specific information.
Packaging: Package of 20.
Customers Who Bought This Also Bought