ContactUsLinkComponent
PIG® Straight-Sided Open-Head UN Rated Poly Drum, New Pig
PIG® Straight-Sided Open-Head UN Rated Poly Drum, New Pig
# de catalogue 10860-600
Fournisseur:  New Pig
UN Rated Poly Drum
PIG® Straight-Sided Open-Head UN Rated Poly Drum, New Pig
# de catalogue 10860-600
Fournisseur:  New Pig
Spécifications
  • Description:
    Straight-sided open-head UN rated poly drum, no bung holes
  • Cat. no.:
    10860-600
  • Capacity:
    55 gal.
  • Dimensions:
    23Ø×37"H
  • Supplier No.:
    DRM1059
Spécifications
A propos de cet article
Straight sidewall, UN rated open top drum has ribs for extra stength and allows broader use of drum grabs and other equipment.
  • Straight sidewalls allow broader use of drum grabs, parrot beak lifters and other equipment that can't move tapered drums
  • Ribs add strength and rigidity, UN Rated for storage and transport of solid hazardous materials
  • High-density polyethylene won't rust or corrode
  • Quick-release lever lock band can be wire sealed to deter unauthorized access
Compatible with a wide range of liquids and chemicals.
Customers Who Bought This Also Bought