A propos de cet article
VWR®/VFH-06/19-9 Vacuum Pump Storage Fume Hood Cabinets are designed without a bottom to allow vacuum pumps and other equipment to be rolled in or out of the cabinets.
- 20" deep
- Right/left
The interior is lined with 1" thick neoprene foam for sound deadening and easy cleaning. Each cabinet is furnished with a 120 VAC, 20 amp, duplex receptacle mounted on the inside cabinet back and a pilot lighted toggle switch mounted in the top front rail. (Wiring is not included.) Each cabinet is furnished with a 1½" I.D. flexible polyolefin tube for venting to the fume hood above. (Requires a 2" hole in the hood work top.) The toespace rail is attached to the door to allow total access to the cabinet. Cabinet inside clearance at the floor is 14½" front-to-back, 25" high, and 3" less than the overall cabinet length.