VWR® Contour™ Acid Storage Fume Hood Base Cabinets
Fournisseur: VWR International
Certificats
A propos de cet article
VWR®/VFH-06/19-8 Steel cabinets (35" high) acid storage fume hood base cabinets are specifically designed for the storage of corrosive chemicals.
- 20" deep
- Lined unit with vent
Available in either steel or wood. These cabinets are lined with a molded one piece linear low density polyethylene tub with coved corners and a 1" lip at the bottom front. The cabinet doors are lined with ⅛" sheet polyethylene and the doors are latched using a nylon roller catch. Each cabinet is furnished with a 1½" I.D. flexible polyolefin tube for venting to the fume hood above. (Requires a 2" hole in the hood work top.)
Customers Who Bought This Also Bought