VWR® Contour™ Fume Hood Base Cabinets
Fournisseur: VWR International
Certificats
A propos de cet article
VWR® Venturi fume hood base cabinet.
VWR®/VFH-06/19-6 Steel cabinets (35" high) standing height steel and wood fume hood base cabinets are 35" high and a nominal 20" deep. ADA height steel fume hood base cabinets are 32" high and 20" deep; ADA height wood fume hood base cabinets are 32½" high and nominal 20" deep. Lengths as shown.
Customers Who Bought This Also Bought