Floor Stripe™, High Performance Message Strips, Accuform
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
A propos de cet article
Floor Stripe™ high performance message strips alert and remind those in and around walkways, work areas and entrances with bold graphics and messages.
- Superior durability for marking in high-traffic locations
- Constructed of low profile, polyester tape with high gloss overlaminate
- Acrylic adhesive backing provides high tack bond
- Apply to clean, non-coated surfaces, including concrete
Customers Who Bought This Also Bought