A propos de cet article
RNA-Seq made simple.
- The fastest and easiest total RNA-seq library kit: Prepare stranded, ribofree libraries from total RNA in 3.5 hours
- Compatible with any organism: Probe-free technology depletes rRNA and globin from any RNA source
- The most accurate: Eliminate bias from rRNA depletion
Discover the most streamlined, stranded RNA library kit for sequencing total RNA from any biological sample with the Zymo-Seq RiboFree™ total RNA library kit. Capture any sample's full transcriptome (both coding and non-coding) with one simple RNA library prep kit. Also capture long-noncoding RNAs (lncRNA), intronic RNAs, nucleolar RNAs, and mRNAs with degraded poly(A) tails.
Ribosomal RNAs (rRNA) comprise approximately 90% of the total RNA and represent an obstacle to transcriptome enrichment of unique protein coding sequences. Zymo Research's RiboFree™ universal depletion effectively removes rRNA and overrepresented transcripts (e.g. globin). Without the use of probes that cause off-target digestion compared to popular rRNA removal kits.