Production and Quality Control Tags, Accuform
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
A propos de cet article
Production and quality control tags support 5S lean programs by helping to identify inspected or defective products.
Material options:
PF-Cardstock - thin, flexible, poly-based cardstock without wood content. They are durable, tear-resistant, waterproof, and will not crack or shrink, when compared to traditional paper cardstock.
RP-Plastic - strong and durable plastic resists chemicals, tears, and abrasions, plus withstands dirt and grease. Top has ³/₈" hole reinforced with metal grommet to prevent pull-off up to 50 lbs. strength.
Production and quality control tags, INSPECTED/TO BE INSPECTED and RELEASED/HOLD: Perforated, two-part tag eliminates the need to re-tag when the status changes.
Customers Who Bought This Also Bought